Order Entry
Germany
ContactUsLinkComponent
2,3-Dichlorobiphenyl (PCB No. 5)
2,3-Dichlorobiphenyl (PCB No. 5)
Catalog # EHERC20000500
Supplier:  EHRENSTORFER
CAS Number:  
2,3-Dichlorobiphenyl (PCB No. 5)
Catalog # EHERC20000500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :223.98 g/mol
UN:3432
ADR:9,II
CAS Number:16605-91-7
Formula:C₁₂H₈Cl₂
MDL Number:MFCD00055244