2,3-Dichlorobiphenyl (PCB No. 5)
Catalog # PROOCIL-PCB-5-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:223.98 g/mol
UN:3432
ADR:9,II
CAS Number:16605-91-7
Formula:C₁₂H₈Cl₂
MDL Number:MFCD00055244