2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB No. 189)
Catalog # PROOCIL-PCB-189-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
ADR:9,II
:395.32 g/mol
UN:3432
CAS Number:39635-31-9
Formula:C₁₂H₃Cl₇