


4-Bromobiphenyl (PBB No. 3) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92-66-0
:
Formula:C₁₂H₉Br
MDL Number:MFCD00000100
:233.11 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:89…92 °C
Density:1,615 g/cm³ (20 °C)
Flash Pt:143 °C
EINECS:202-176-6
UN:3077
:GHS07
:GHS05
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline flakes
- Infrared spectrum:Conforms
- Melting point:89 to 92 °C
- GC:≥98.5 %