Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Fluoromesitylene
2-Fluoromesitylene 98%
2-Fluoromesitylene 98%
:
Thermo Fisher Scientific
2-Fluoromesitylene 98%
:
Thermo Fisher Scientific
:
392-69-8
:
:
C₉H₁₁F
:
MFCD00052365
:
163…165 °C
:
0.975
:
138.18 g/mol
:
GHS07
:
Ambient
msds_ALFAA12779
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years