Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Di-tert-butylhydroquinone
2,5-Di-tert-butylhydroquinone ≥98%
2,5-Di-tert-butylhydroquinone ≥98%
:
Thermo Fisher Scientific
2,5-Di-tert-butylhydroquinone ≥98%
:
Thermo Fisher Scientific
:
88-58-4
:
:
C₁₄H₂₂O₂
:
MFCD00008825
:
222.33 g/mol
:
321 °C
:
214…218 °C
:
1.070
:
>200 °C (392 °F)
:
Ambient
:
201-841-8
:
GHS06
:
GHS07
msds_ALFAA14606
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years