Order Entry
Germany
ContactUsLinkComponent
Supelco® Organic Reference Standard, Thymol
Supelco® Organic Reference Standard, Thymol
Catalog # 72477-500MG-F
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Thymol
Catalog # 72477-500MG-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    500 mg
  • Standards:
    Analytical standard
  • CAS No.:
    89-83-8
  • Description:
    Thymol
  • Storage conditions:
    RT
  • Hazard class description:
    H302 - H314 - H411
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00002309
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-Isopropyl-5-methylphenol; 5-Methyl-2-(1-methylethyl)phenol; 5-Methyl-2-isopropylphenol; IPMP
  • Melting point:
    48 - 51 °C (lit.); 49 - 51 °C
  • Formula:
    2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH
  • Molecular weight:
    150,22
  • Density:
    0,965 g/ml at 25 °C (lit.)
  • Flash point:
    101 °C - Closed cup
  • Boiling point:
    232 °C (lit.)

Specifications

About this item

  • GHS signal: GHS05, GHS07, GHS09
Caution: P260 - P273 - P280 - P301 + P312 - P303 + P361 + P353 - P305 + P351 + P338