


2,6-Xylenol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:576-26-1
:
Formula:C₈H₁₀O
MDL Number:MFCD00002240
:122.17 g/mol
Merck Index:12,10215
Boiling Pt:203 °C (1013 hPa)
Melting Pt:41…42 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:73 °C
Storage Temperature:Ambient
EINECS:209-400-1
UN:2261
ADR:6.1,II
:GHS06
:GHS05
:GHS09