4-Iodo-o-xylene
Catalog # FLUO004948-25G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:232.06 g/mol
Boiling Pt:231…233 °C (1013 hPa)
Density:1,633 g/cm³ (20 °C)
:GHS07
CAS Number:31599-61-8
Formula:C₈H₉I
MDL Number:MFCD00040989