

3,5-Bis(trifluoromethyl)phenyl isothiocyanate 99+%
:23165-29-9
:
:C₉H₃F₆NS
:MFCD00040838
:271.19 g/mol
:63 °C (1.5 mmHg)
:1.485 g/cm³
:Ambient
:1760
:8,III
:GHS07
:GHS05
- Appearance:Clear to slightly hazy colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4980 to 1.5020 (20°C, 589 nm)
- GC:≥99.0 %