Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-tert-Butyl-α-chlorotoluene
4-tert-Butyl-α-chlorotoluene ≥99%
4-tert-Butyl-α-chlorotoluene ≥99%
:
Thermo Fisher Scientific
4-tert-Butyl-α-chlorotoluene ≥99%
:
Thermo Fisher Scientific
:
19692-45-6
:
:
C₁₁H₁₅Cl
:
MFCD00000918
:
182.69 g/mol
:
101…104 °C (7 mmHg)
:
0.945
:
94° (202 °F)
:
Ambient
:
243-228-8
:
1760
:
8,III
:
GHS05
msds_ALFAA11856
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years