Order Entry
Germany
ContactUsLinkComponent
4-Methoxybenzophenone 97%
4-Methoxybenzophenone 97%
Certificates
undefined
4-Methoxybenzophenone 97%

Specification Test Results

  • Appearance:
    White to yellow to light orange Crystalline solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    58 to 63 °C
  • GC:
    ≥96.0 %

Specification Test Results

Related Information