Order Entry
Germany
ContactUsLinkComponent
Bromocresol purple
Bromocresol purple
Catalog # MOLE46156907-25G
Supplier:  Molekula
CAS Number:  
Bromocresol purple
Catalog # MOLE46156907-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :540.23 g/mol
Merck Index:13,01371
Melting Pt:242 °C
EINECS:204-087-8
 :GHS07
Storage Temperature:Ambient
CAS Number:115-40-2
Formula:C₂₁H₁₆Br₂O₅S
MDL Number:MFCD00011681