Order Entry
Germany
Orders LinkContactUsLinkComponent
Bromophenol blue

Bromophenol blue

Bromophenol blue
Certificates
Bromophenol blue
Bromophenol blue

2 Options Available Below

Product Details & Documents

CAS No.: 115-39-9 Molecular Formula: C19H10Br4O5S Formula Weight: 669.98 Odour: Odourless Melting Point: ca 270° dec. Beilstein Reference: 61698 Merck Reference: 14,1444 MDL No.: MFCD00005875

Synonyms:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS Number:115-39-9
  • Formula:C₁₉H₁₀Br₄O₅S
  • MDL Number:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:14,1444
  • Melting Pt:∼ 270 °C (dec.)
  • Density:1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-086-2
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.