Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Propylbenzene
Propylbenzene, Sigma-Aldrich®
Propylbenzene, Sigma-Aldrich®
:
Merck
Propylbenzene, Sigma-Aldrich®
:
Merck
:
103-65-1
:
:
C₉H₁₂
:
MFCD00009377
:
120.19 g/mol
:
12,08029
:
159 °C (1013 hPa)
:
–99 °C
:
0.862 g/cm³ (20 °C)
:
45 °C
:
203-132-9
:
2364
:
3,III
:
GHS02
:
GHS08
:
GHS07
:
GHS09
:
Ambient