Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
o-Toluenesulphonyl chloride
o-Toluenesulphonyl chloride 80%
o-Toluenesulphonyl chloride 80%
:
Thermo Fisher Scientific
o-Toluenesulphonyl chloride 80%
:
Thermo Fisher Scientific
:
133-59-5
:
:
C₇H₇ClO₂S
:
MFCD00024874
:
190.65 g/mol
:
154 °C (10 mmHg)
:
10 °C
:
1.338
:
>110 °C (230 °F)
:
Ambient
:
205-113-0
:
3265
:
8,II
:
GHS05
msds_ALFAL18983
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years