Order Entry
Germany
ContactUsLinkComponent
4-Ethylresorcinol, Sigma-Aldrich®
Irritant
4-Ethylresorcinol, Sigma-Aldrich®
Catalog # SIALE48200-25G
Supplier:  Merck
CAS Number:  
Irritant
4-Ethylresorcinol, Sigma-Aldrich®
Catalog # SIALE48200-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :138.17 g/mol
Boiling Pt:283 °C (1013 hPa)
Melting Pt:100 °C
 :M-Clarity™ quality level = MQ100
EINECS:220-777-1
 :GHS07
CAS Number:2896-60-8
Formula:C₈H₁₀O₂
MDL Number:MFCD00002283