2-Phenylethanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:60-12-8
:
Formula:C₆H₅CH₂CH₂OH
MDL Number:MFCD00002886
:122.17 g/mol
Merck Index:12,07370
Boiling Pt:432 °C (1013 hPa)
Melting Pt:103…105 °C
Density:1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-456-2
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ300
Flash Pt:102 °C
:GHS06