Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2-Methoxy-p-cresol

Glass bottle for solids, 100 g
Catalog # MOLE25783219-100G
Irritant
Marketsource
Supplier:   Molekula
2-Methoxy-p-cresol
Catalog # MOLE25783219-100G
Supplier:  Molekula
Marketsource
CAS Number:  93-51-6
2-Methoxy-p-cresol

Glass bottle for solids, 100 g

CAS Number:  93-51-6
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Creosol, 1-Hydroxy-2-methoxy-4-methylbenzene, 2-Methoxy-p-cresol, 2-Methoxy-4-methylphenol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, 2-Hydroxy-5-methylanisol
  • CAS Number:93-51-6
  • Formula:C₈H₁₀O₂
  • MDL Number:MFCD00002378
  • Molecular Weight:138.17 g/mol
  • Merck Index:13,02599
  • Boiling Pt:219…222 °C (1013 hPa)
  • Density:1.09 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:202-252-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications