Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
N-Ethyl-o-toluidine
N-Ethyl-o-toluidine for synthesis, Sigma-Aldrich®
N-Ethyl-o-toluidine for synthesis, Sigma-Aldrich®
:
Merck
N-Ethyl-o-toluidine for synthesis, Sigma-Aldrich®
:
Merck
:
94-68-8
:
:
C₉H₁₃N
:
MFCD00026695
:
135.21 g/mol
:
212…212.5 °C (1013 hPa)
:
–15 °C
:
0.95 g/cm³ (20 °C)
:
Ambient
:
202-354-3
:
6.1,II
:
M-Clarity™ quality level = MQ200
:
2754
msds_843602