4-Aminobenzyl alcohol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:623-04-1
:
Formula:C₇H₉NO
MDL Number:MFCD00014782
:123.15 g/mol
Boiling Pt:169…171 °C (11 mmHg)
Melting Pt:75 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:210-767-5