Order Entry
Germany
ContactUsLinkComponent
2-Aminobenzyl alcohol
2-Aminobenzyl alcohol
Catalog # MOLE13182142-25G
Supplier:  Molekula
CAS Number:  
2-Aminobenzyl alcohol
Catalog # MOLE13182142-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :123.15 g/mol
Boiling Pt:275 °C (1013 hPa)
Melting Pt:81 °C
Storage Temperature:Refrigerator
EINECS:226-293-7
CAS Number:5344-90-1
Formula:C₇H₉NO
MDL Number:MFCD00007749