
2-Aminobenzyl alcohol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5344-90-1
:
Formula:C₇H₉NO
MDL Number:MFCD00007749
:123.15 g/mol
Boiling Pt:162 °C (15 mmHg)
Melting Pt:82…86 °C
Storage Temperature:Ambient
EINECS:226-293-7
:GHS07
Specification Test Results
- Appearance:Beige to grey-brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:82 to 86 °C
- GC:≥97.5 %