Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Trifluoromethyl)thiophenol
4-(Trifluoromethyl)thiophenol
4-(Trifluoromethyl)thiophenol
4-(Trifluoromethyl)thiophenol
Supplier:
FLUOROCHEM
Certificates
Expand image
4-(Trifluoromethyl)thiophenol
Supplier:
FLUOROCHEM
About this item
CAS Number:
825-83-2
:
Formula:
C₇H₅F₃S
MDL Number:
MFCD00042320
:
178.18 g/mol
Boiling Pt:
60…61 °C (13 mmHg)
Density:
1.31 g/cm³ (25 °C)
UN:
2810
ADR:
6.1,III