Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Trifluoromethyl)phenyl isothiocyanate
2-(Trifluoromethyl)phenyl isothiocyanate
2-(Trifluoromethyl)phenyl isothiocyanate
:
FLUOROCHEM
2-(Trifluoromethyl)phenyl isothiocyanate
:
FLUOROCHEM
:
1743-86-8
:
:
C₈H₄F₃NS
:
MFCD00039644
:
203.19 g/mol
:
214…216 °C (1013 hPa)
:
1,346 g/cm³ (20 °C)