Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Trifluoromethyl)phenyl isocyanate
4-(Trifluoromethyl)phenyl isocyanate
4-(Trifluoromethyl)phenyl isocyanate
:
Thermo Scientific
4-(Trifluoromethyl)phenyl isocyanate
:
Thermo Scientific
:
1548-13-6
:
:
C₈H₄F₃NO
:
MFCD00002028
:
187.12 g/mol
:
173 °C (1013 hPa)
:
58…59 °C
:
1.31 g/cm³ (20 °C)
:
Refrigerator
:
216-284-6
:
2285
:
6.1,II