2'-Fluoroacetophenone
Glass bottle for solids, 25 g
Catalog # MOLE29373603-25G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 1-(2-Fluorophenyl)ethanone, o-Fluoroacetophenone
- CAS Number:445-27-2
- Formula:C₈H₇FO
- MDL Number:MFCD00000320
- Molecular Weight:138.14 g/mol
- Boiling Pt:82…83 °C (13,3 hPa)
- Boiling Pt:82…83 °C (13.3 hPa)
- Density:1,12 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-156-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



