2'-Fluoroacetophenone 97%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
2'-Fluoroacetophenone 97%
Catalog # ACRO179780250
Supplier: Thermo Fisher Scientific
CAS Number: 445-27-2
Jump to:
Product Details & Documents
Synonyms: 1-(2-Fluorophenyl)ethanone, o-Fluoroacetophenone
- CAS Number:445-27-2
- Formula:C₈H₇FO
- MDL Number:MFCD00000320
- Molecular Weight:138.14 g/mol
- Boiling Pt:82…83 °C (10 mmHg)
- Density:1.137 g/cm³
- Flash Pt:61 °C
- Storage Temperature:Ambient
- EINECS:207-156-0
Documents
ACRO17978
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5065 to 1.5085 (20°C, 589 nm)
- GC:≥96.0 %
- Titration with HClO4:99 to 101 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



