Benzotrifluoride
Glass bottle for solids, 100 mg
Catalog # PROOMM0161.09
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
Synonyms: α,α,α-Trifluorotoluene, Solvent BTF
- CAS Number:98-08-8
- Formula:C₇H₅F₃
- MDL Number:MFCD00000372
- Molecular Weight:146.11 g/mol
- Merck Index:13,01111
- Boiling Pt:101 °C (1013 hPa)
- Melting Pt:–29 °C
- Density:1.37 g/cm³ (20 °C)
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:202-635-0
- UN:2338
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



