Order Entry
Germany
ContactUsLinkComponent
 
 
 
4-tert-Butylbenzoyl chloride 98%
 
undefined
 
 
4-tert-Butylbenzoyl chloride 98%

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5354 to 1.5374 (20°C, 589 nm)
  • Titration RCOCl:
    ≥97.5 %