4'-Methoxyacetophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-06-1
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00008745
:150.18 g/mol
Boiling Pt:152 °C (1013 hPa)
Melting Pt:36…38 °C
Density:1.14 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:202-815-9
:GHS07
Storage Temperature:Ambient