4'-Methoxyacetophenone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-06-1
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00008745
:150.18 g/mol
Boiling Pt:152 °C (1013 hPa)
Melting Pt:36…39 °C
Density:1,14 g/cm³ (20 °C)
Flash Pt:138 °C
EINECS:202-815-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline mass
- Infrared spectrum:Conforms
- Melting point:36 to 40 °C
- GC:≥97.5 %