4'-Ethoxyacetophenone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1676-63-7
:
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00009095
:164.2 g/mol
Boiling Pt:268…269 °C (758 mmHg)
Melting Pt:35…38 °C
Flash Pt:134 °C
Storage Temperature:Ambient
EINECS:216-825-6
:GHS07
Specification Test Results
- Appearance:White to yellow Crystals and/or chunks or low melting solid
- Infrared spectrum:Conforms
- Melting point:35 to 38 °C
- GC:≥98.5 %