Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2'-Methoxyacetophenone ≥98%

2'-Methoxyacetophenone ≥98%
Marketsource
Certificates
2'-Methoxyacetophenone ≥98%
2'-Methoxyacetophenone ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 579-74-8 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 108°(226°F) Refractive Index: 1.5393 Boiling Point: 119-120°/10mm Beilstein Reference: 1100602 MDL No.: MFCD00008725

Synonyms:  1-(2-Methoxyphenyl)ethanone
  • CAS Number:579-74-8
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00008725
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:119…120 °C (10 mmHg)
  • Density:1.090
  • Flash Pt:108 °C (226 °F)
  • Storage Temperature:Ambient
  • EINECS:209-446-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 46,80 €
    46,80 €
    1 * 25 g
       46,80 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 144,00 €
    144,00 €
    1 * 100 g
       144,00 €