Order Entry
Germany
Orders LinkContactUsLinkComponent
2'-Methoxyacetophenone 99%

2'-Methoxyacetophenone 99%

Irritant
2'-Methoxyacetophenone 99%
Certificates
2'-Methoxyacetophenone 99%

2 Options Available Below

Product Details & Documents
Synonyms:  1-(2-Methoxyphenyl)ethanone
  • CAS Number:579-74-8
  • Formula:C₉H₁₀O₂
  • MDL Number:MFCD00008725
  • Molecular Weight:150.18 g/mol
  • Boiling Pt:131 °C (18 mmHg)
  • Density:1.09 g/cm³
  • Flash Pt:108 °C
  • Storage Temperature:Ambient
  • EINECS:209-446-2

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear light yellow to orange to light brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5383 to 1.5403 (20°C, 589 nm)
  • GC:
    ≥98.5 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 48,60 €
    48,60 €
    1 * 25 mL
       48,60 €

  • Glass bottle
    100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - 129,00 €
    129,00 €
    1 * 100 mL
       129,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.