4-Acetylbenzoic acid
Catalog # PROOMM0002.08
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:164.16 g/mol
Boiling Pt:340 °C (1013 hPa)
EINECS:209-588-5
CAS Number:586-89-0
Formula:C₉H₈O₃
MDL Number:MFCD00002561