Dodecanophenone, Sigma-Aldrich®
Catalog # SIAL252719-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:260.42 g/mol
Boiling Pt:148…152 °C (0.6 mmHg)
Melting Pt:44…46 °C
Density:0.9045 g/cm³ (41.5 °C)
:M-Clarity™ quality level = MQ100
EINECS:216-818-8
CAS Number:1674-38-0
Formula:C₆H₅C(O)(CH₂)₁₀CH₃
MDL Number:MFCD00008967