Order Entry
Germany
ContactUsLinkComponent
2-Chloro-5-nitrobenzyl alcohol
2-Chloro-5-nitrobenzyl alcohol
Catalog # MOLE29814067-5G
Supplier:  Molekula
CAS Number:  
2-Chloro-5-nitrobenzyl alcohol
Catalog # MOLE29814067-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :187.58 g/mol
Melting Pt:78 °C
EINECS:279-584-6
CAS Number:80866-80-4
Formula:C₇H₆ClNO₃
MDL Number:MFCD00007297