Order Entry
Germany
ContactUsLinkComponent
 
 
 
5-Chloro-o-toluidine for synthesis, Sigma-Aldrich®
  8.02641.0250
 :  Merck
 :  
 
 
5-Chloro-o-toluidine for synthesis, Sigma-Aldrich®
  8.02641.0250
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Seq:
    2590

 

 

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    22 °C
  • Molar Mass:
    141.59 g/mol
  • Boiling Point:
    237 °C (1013 hPa)
  • Vapor Pressure:
    0.45 hPa (50 °C)
  • Flash Point:
    160 °C
  • Refractive Index:
    1.584 (20 °C, 589 nm)
  • Density:
    1.17 g/cm3 (20 °C)
  • Ignition Point:
    540 °C

 

 

 :
 :141.6 g/mol
 :237 °C (1013 hPa)
 :22 °C
 :1.17 g/cm³ (20 °C)
 :160 °C
 :Ambient
 :202-452-6
 :3429
 :6.1,III
 :M-Clarity™ quality level = MQ200
 :GHS08
 :GHS07
 :95-79-4
 :C₇H₈ClN
 :MFCD00007779