2,4-Dichloro-3,5-dinitro-α,α,α-trifluorotoluene
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:29091-09-6
:
Formula:C₇HCl₂F₃N₂O₄
MDL Number:MFCD00042480
:305 g/mol
Boiling Pt:291…294 °C (1013 hPa)
Melting Pt:74…75 °C
EINECS:249-420-8
UN:3077
ADR:9,III