
6-Chloro-m-toluic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6342-60-5
:
Formula:C₈H₇ClO₂
MDL Number:MFCD00045798
:170.6 g/mol
Melting Pt:148…151 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to beige Powder
- Infrared spectrum:Conforms
- Melting point:148 to 151 °C
- HPLC:≥97.5 %