ω-Bromo-2-methoxyacetophenone
Catalog # MOLE35811823-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:229.07 g/mol
Boiling Pt:324 °C (1013 hPa)
Melting Pt:40…44 °C
Storage Temperature:Refrigerator
EINECS:250-870-2
UN:3261
ADR:8,III
CAS Number:31949-21-0
Formula:C₉H₉BrO₂
MDL Number:MFCD00000196