ω,4-Dibromoacetophenone
Marketsource
Supplier: FLUOROCHEM
Jump to:
About this item
Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS Number:99-73-0
- Formula:C₈H₆Br₂O
- MDL Number:MFCD00000200
- Molecular Weight:277.94 g/mol
- Merck Index:13,01412
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:107…108 °C
- Storage Temperature:Refrigerator
- EINECS:202-783-6
- UN:3261
- ADR:8,II
2 Options Available Below
Product Details & Documents
Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS Number:99-73-0
- Formula:C₈H₆Br₂O
- MDL Number:MFCD00000200
- Molecular Weight:277.94 g/mol
- Merck Index:13,01412
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:107…108 °C
- Storage Temperature:Refrigerator
- EINECS:202-783-6
- UN:3261
- ADR:8,II
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



