Order Entry
Germany
Orders LinkContactUsLinkComponent
 

ω,4-Dibromoacetophenone ≥98%

ω,4-Dibromoacetophenone ≥98%
Certificates
ω,4-Dibromoacetophenone ≥98%
ω,4-Dibromoacetophenone ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 99-73-0 Molecular Formula: C8H6Br2O Formula Weight: 277.95 Storage Temperature: 2°C to 8°C Odour: Pungent Beilstein Reference: 607604 Merck Reference: 14,1427 UN No.: 3261 MDL No.: MFCD00000200

Synonyms:  Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
  • CAS Number:99-73-0
  • Formula:C₈H₆Br₂O
  • MDL Number:MFCD00000200
  • Molecular Weight:277.94 g/mol
  • Merck Index:14,1427
  • Boiling Pt:310 °C (1013 hPa)
  • Melting Pt:108…112 °C
  • Storage Temperature:Refrigerator
  • EINECS:202-783-6
  • UN:3261
  • ADR:8,II
Product Family Options

Product Information