ω,4-Dibromoacetophenone 98%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS Number:99-73-0
- Formula:C₈H₆Br₂O
- MDL Number:MFCD00000200
- Molecular Weight:277.94 g/mol
- Merck Index:13,01412
- Boiling Pt:1415 °C (760 mmHg)
- Melting Pt:107…111 °C
- Storage Temperature:Ambient
- EINECS:202-783-6
- UN:3261
- ADR:8,II
3 Options Available Below
Product Details & Documents
Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS Number:99-73-0
- Formula:C₈H₆Br₂O
- MDL Number:MFCD00000200
- Molecular Weight:277.94 g/mol
- Merck Index:13,01412
- Boiling Pt:1415 °C (760 mmHg)
- Melting Pt:107…111 °C
- Storage Temperature:Ambient
- EINECS:202-783-6
- UN:3261
- ADR:8,II
Specifications for Product Family
Specification Test Results
- Appearance:Light yellow to beige Crystalline solid
- Infrared spectrum:Conforms
- Melting point:107 to 111 °C
- GC:≥97.5 %



