(2-Bromophenyl)acetonitrile
Marketsource
Supplier: FLUOROCHEM
Jump to:
About this item
Synonyms: 2-Bromobenzyl cyanide
- CAS Number:19472-74-3
- Formula:C₈H₆BrN
- MDL Number:MFCD00001896
- Molecular Weight:196.05 g/mol
- Boiling Pt:270 °C (1013 hPa)
- Melting Pt:1 °C
- Density:1.51 g/cm³ (20 °C)
- EINECS:243-091-4
- UN:1694
- ADR:6.1,I
- :Density: 1.51 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
Synonyms: 2-Bromobenzyl cyanide
- CAS Number:19472-74-3
- Formula:C₈H₆BrN
- MDL Number:MFCD00001896
- Molecular Weight:196.05 g/mol
- Boiling Pt:270 °C (1013 hPa)
- Melting Pt:1 °C
- Density:1.51 g/cm³ (20 °C)
- EINECS:243-091-4
- UN:1694
- ADR:6.1,I
- :Density: 1.51 g/cm³ (20 °C)
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



