Order Entry
Germany
ContactUsLinkComponent
Resveratrol, Supelco®
Resveratrol, Supelco®
Catalog # 34092-100MG
Supplier:  Merck
CAS Number:  
undefined
Resveratrol, Supelco®
Catalog # 34092-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :228.25 g/mol
Merck Index:12,08326
Melting Pt:260 °C
Density:1.36 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Freezer
CAS Number:501-36-0
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00133799