Order Entry
Germany
ContactUsLinkComponent
 
Resveratrol, Millipore®
Catalog # 554325-25
CAS Number:  
Resveratrol, Millipore®
Catalog # 554325-25
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :228.25 g/mol
Merck Index:12,08326
Melting Pt:260 °C
Density:1.36 g/cm³ (20 °C)
Storage Temperature:Freezer
CAS Number:501-36-0
Formula:C₁₄H₁₂O₃
MDL Number:MFCD00133799