Order Entry
Germany
ContactUsLinkComponent
Styrene
Styrene
Catalog # PROOCHE140
CAS Number:  
Styrene
Catalog # PROOCHE140
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2 mL
  • Seq:
    2145

Specifications

About this item

 :
 :104.15 g/mol
Merck Index:13,08944
Boiling Pt:145.2 °C (1013 hPa)
Melting Pt:–31 °C
Density:0.906 g/cm³ (20 °C)
Flash Pt:31 °C
EINECS:202-851-5
UN:2055
ADR:3,III
 :GHS02
 :GHS07
CAS Number:100-42-5
Formula:C₈H₈
MDL Number:MFCD00008612