Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Styrene
Styrene
Styrene
Styrene
KRAE30-120.5000
:
KRAEMER MARTIN
:
Styrene
KRAE30-120.5000
:
KRAEMER MARTIN
:
Pk:
5 L
Seq:
5190
:
:
104.15 g/mol
:
13,08944
:
145.2 °C (1013 hPa)
:
–31 °C
:
0.906 g/cm³ (20 °C)
:
31 °C
:
202-851-5
:
2055
:
3,III
:
GHS02
:
GHS07
:
100-42-5
:
C₈H₈
:
MFCD00008612